Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ADP Assay Kit

Catalog No. 89028390 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-390 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-390 Supplier Abnova™ Supplier No. KA1658
Add to Cart
Add to Cart

Rapid bioluminescent determination of ADP in biological sample and cell

Specifications

Content And Storage Store the kit at -20°C.
Description ADP Assay Kit is a rapid bioluminescent determination of ADP.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 120μL substrate, 120μL Co substrate, 120μL ATP enzyme, 120μL ADP enzyme, 100μL standard 3mM ADP
Quantity 1 Kit
Sample Type Biological Sample, Cell
Sample Volume 100 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.