Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ADP/ATP Ratio Assay Kit

Catalog No. 89028405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-405 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-405 Supplier Abnova™ Supplier No. KA1673
Add to Cart
Add to Cart

Bioluminescent assay for ADP/ATP ratio

  • Sample Type: Cell

Functional Study

Specifications

Content And Storage Store the kit at -20°C.
Description ADP/ATP Ratio Assay Kit is a bioluminescent assay for ADP/ATP ratio.
For Use With (Application) Functional Study
Includes 10mL assay buffer, 120μL substrate, 120μL Co substrate, 120μL ATP enzyme, 120μL ADP enzyme
Quantity 1 Kit
Sample Type Cell
Type ADP/ATP Ratio Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.