Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ AF HDL and LDL/VLDL Assay Kit

Catalog No. 89028388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-388 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-388 Supplier Abnova™ Supplier No. KA1656
Add to Cart
Add to Cart

Used for quantitative colorimetric/fluorimetric determination of HDL and LDL/VLDL

  • Sample Type: Serum
  • Calibration Range: Colorimetric Assay: 1 to 100mg/dL, Fluorimetric Assay: 0.2 to 10mg/dL

Functional Study

Specifications

Calibration Range 1 to 100mg/dL colorimetric assay, 0.2 to 10mg/dL fluorimetric assay
Content And Storage Store the kit at -20°C. Avoid repeated freezing and thawing.
Description AF HDL and LDL/VLDL Assay Kit is a quantitative colorimetric/fluorimetric determination of HDL and LDL/VLDL.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 1.5mL x 2 PBS, 20mL assay buffer, 120μL dye reagent, 1.5mL precipitation reagent, 120μL enzyme mix, 1mL 300mg/dL Cholesterol standard
Quantity 1 Kit
Sample Type Serum
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.