Learn More
Description
Includes
- ALT Assay Buffer (10X), 1 vial
- ALT Substrate, 1 vial
- ALT Cofactor, 2 vials
- ALT Initiator, 1 vial
- ALT Positive control, 1 vial
- 96-Well Plate (colorimetric assay),1 plate
- 96-Well cover sheet,1 cover
- Detect ALT activity in serum, plasma, tissue samples, and cell lysates
- Measurement of the ALT activity is carried out by monitoring the rate of NADH oxidation in a coupled reaction system employing lactate dehydrogenase (LDH)
Specifications
Specifications
| Content And Storage | Store the kit at -80°C. |
| Description | Alanine Transaminase Assay Kit is a colorimetric method for assaying the activity of ALT. |
| For Use With (Application) | Flow Cytometry, Functional Study |
| Includes | 1 vial ALT assay buffer 10X, 1 vial ALT substrate, 2 vials ALT Co-factor, 1 vial ALT initiator, 1 vial ALT positive control, 1 plate 96-well plate (colorimetric assay), 1 cover 96-well cover sheet |
| Quantity | 1 Kit |
| Type | Activity Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.