Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Pig) ELISA Kit

Catalog No. 89028163 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-163 Plasma, Serum
89-100-226 CCS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-163 Supplier Abnova™ Supplier No. KA1027
Add to Cart
Add to Cart

Competitive Enzyme Immunoassay for the quantitative measurement of pig ALB in Plasma and Serum

Specifications

Calibration Range 1.56 to 100μg/mL
Conjugate HRP
Cross Reactivity Pig
Description ALB (Pig) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of pig ALB.
Detection Limit 0.3μg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 200μg swine Albumin standard, 1 vial Biotinylated Albumin, 30mL mix diluent concentrate (10X), 30mL Wash Buffer 20X, 90 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL st
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Albumin (Pig) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.