Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Pig) ELISA Kit

Catalog No. 89100226 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-226 CCS
89-028-163 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-226 Supplier Abnova™ Supplier No. KA0497
Add to Cart
Add to Cart

Competitive Enzyme Immunoassay for the quantitative measurement of pig ALB in Plasma and Serum

Specifications

Calibration Range 1.56 to 100ng/mL
Conjugate Biotin
Cross Reactivity Pig
Description Albumin (Pig) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of pig albumin.
Detection Limit 1ng/mL
Detection Method Colorimetric
Includes 96 wells-antibody coated microwells, Swine albumin standard (3.2ug), Biotinylated Swine albumin antibody (1vial), MIX Diluent concentrate 10X (30mL), Wash Buffer Concentrate 20X (2 x 30mL), 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Albumin (Pig) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.