Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Rat), HRP, ELISA Kit

Catalog No. 89100681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-681 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-681 Supplier Abnova™ Supplier No. KA2491
Add to Cart
Add to Cart

Includes

  • anti-rat albumin-coated wells 96 wells (8 x 12 strips)
  • Reference standard Lyophilized
  • 10x Diluent 50mL
  • Enzyme Conjugate Reagent 11mL
  • 20x Wash Solution 50mL
  • TMB Reagent (One step) 11mL
  • Stop Solution (1N HCl) 11mL

For quantitative measurement of TNNI3 in plasma and serum

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Albumin (Rat) ELISA Kit is for the quantitative measurement of Albumin in serum.
Detection Limit 1.56 to 100 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Albumin (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.