Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alkaline Phosphatase Assay Kit

Catalog No. 89028374 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-374 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-374 Supplier Abnova™ Supplier No. KA1642
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Colorimetric kinetic determination of serum alkaline phosphatase activity

  • Sample Volume: 5-50μL
  • Sample Type:: Plasma, Serum

Functional Study

Specifications

Calibration Range 2 to 800U/L
Content And Storage Store pNPP Liquid at -20°C. Store other components at 4°C.
Description Alkaline Phosphatase Assay Kit is a colorimetric kinetic determination of serum alkaline phosphatase activity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL assay buffer (pH 10.5), 1.5mL Mg acetate (0.2M), 600μL pNPP liquid (1M), 10mL calibrator, Tartrazine
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 5 to 50 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.