Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alkaline Phosphatase Assay Kit (Fluorometric)

Catalog No. 89101210 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-210 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-210 Supplier Abnova™ Supplier No. KA3796
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • ALP assay buffer, 100mL
  • MUP substrate, 1 vial
  • ALP enzyme, 1 vial
  • Stop solution, 25mL

To detect alkaline phosphatase (ALP) activity, functional study

Specifications

Content And Storage Store the kit at -20°C.
Description Alkaline Phosphatase Assay Kit is a quantitative fluorimetric determination of alkaline phosphatase at FL360/450 nm.
Detection Method Fluorometric
For Use With (Application) Functional Study
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 10 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.