Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ alpha-Glucosidase Assay Kit

Catalog No. 89028340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-340 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-340 Supplier Abnova™ Supplier No. KA1608
Add to Cart
Add to Cart

Includes

  • Assay buffer: (pH 7.0) 24mL
  • α-NPG substrate: 1mL
  • Calibrator: (equivalent to 250U/L) 10mL

Colorimetric kinetic determination of a-glucosidase activity in biological sample

Specifications

Content And Storage Store the kit at -20°C.
Description alpha-Glucosidase Assay Kit is a colorimetric kinetic determination of a-Glucosidase activity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 24mL assay buffer (pH 7), 1mL a-NPG substrate, 10mL calibrator (equivalent to 250U/L)
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 20 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.