Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ANGPT2 (Mouse) ELISA Kit

Catalog No. 89101103 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-103 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-103 Supplier Abnova™ Supplier No. KA3691
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-mouse Angiopoietin-2 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse Angiopoietin-2 standard, 10ng x 2
  • Biotinylated anti-mouse Angiopoietin-2 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Angpt2. Sandwich enzyme immunoassay.

Angpt2 (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 156 to 10,000pg/mL
Content And Storage Store the kit at -20°C
Description Angpt2 (Mouse) is a sandwich enzyme immunoassay for the quantitative measurement of mouse Angpt2.
Conjugate Biotin
Cross Reactivity Mouse
Detection Method Colorimetric
Product Type Angpt2 (Mouse) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant mouse Angiopoietin-2
Sufficient For 96 Assays
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.