Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Annexin V-FITC/7-AAD Apoptosis Kit (Fluorometric)

Catalog No. 89101292 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-292 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-292 Supplier Abnova™ Supplier No. KA3806
Add to Cart
Add to Cart

Use for apoptosis detection.

Specifications

Content And Storage Store the kit at 4°C.
Description Annexin V-FITC/7-AAD Apoptosis Kit is used for apoptosis detection.
Detection Method Fluorometric
For Use With (Application) Flow Cytometry, Functional Study
Quantity 1 Kit
Sample Type Cell
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.