Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Antioxidant Assay Kit

Catalog No. 89028354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-354 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-354 Supplier Abnova™ Supplier No. KA1622
Add to Cart
Add to Cart

A quantitative colorimetric determination of total antioxidant capacity.

  • Measures total antioxidant capacity in which Cu2+ is reduced by antioxidant to Cu+
  • The resulting Cu+ specifically forms a colored complex with a dye reagent
  • The color intensity at 570nm is proportional to TAC in the sample

Specifications

Calibration Range 1.5 to 1000μM
Content And Storage Store reagent A at room temperature. Store other components at -20°C.
Description Antioxidant Assay Kit is a quantitative colorimetric determination of total antioxidant capacity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 12mL Reagent A, 1mL reagent B,100μL standard 50mM Trolox
Quantity 1 Kit
Sample Type Beverage, Biological Sample, Food, Plasma, Serum, Saliva, Urine
Sample Volume 20 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.