Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Arginase Assay Kit

Catalog No. 89028341 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-341 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-341 Supplier Abnova™ Supplier No. KA1609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Arginine buffer (pH 9.5), 2mL
  • Mn solution, 1mL
  • Reagent A, 25mL
  • Reagent B, 25mL
  • Urea standard (50mg/dL), 1mL

Arginase Assay Kit is a quantitative colorimetric arginase determination.

  • Sensitive, accurate, and convenient: Detection limit of 0.3 U/L for 2-hour arginase reaction in 96-well assay format
  • Simple and high throughput: procedure involves incubation of the provided substrate with the sample in a microplate followed by the addition of the coloring reagent
  • Can be readily automated as a high-throughput assay for thousands of samples per day
  • New procedure increases recovery of small fragmented DNA and therefore improves the sensitivity of the assay
  • Store the Arginine Buffer and Urea Standard at -20°C and other components at 2-8°C; shelf life of 12 months after receipt

Specifications

Content And Storage Store Arginine Buffer and Urea Standard at -20°C. Store other components at 4°C.
Description Arginase Assay Kit is a quantitative colorimetric arginase determination.
For Use With (Application) Functional Study
Includes 2mL Arginine Buffer pH 9.5, 1mL Mn Solution, 25mL reagent A, 25mL reagent B, 1mL Urea Standard (50mg/dL)
Quantity Each
Sample Volume 40μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.