Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -20°C. |
| Description | Ascorbate Assay Kit is a fluorescence-based method for assaying ascorbate. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 2 vials Ascorbate standard, 25mL Ascorbate assay buffer, 500μL Ascorbate DTPA, 2 vials Ascorbate substrate, 2 vials Ascorbate developer, 1 plate 96-well plate (black), 1 cover 96-well cover sheet |
| Quantity | 1 Kit |
| Type | Activity Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.