Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Aspartate Transaminase Assay Kit

Catalog No. 89028357 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-357 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-357 Supplier Abnova™ Supplier No. KA1625
Add to Cart
Add to Cart

Includes

  • Assay buffer, 24mL
  • Cofactors, 120μL
  • Enzyme mix, 120μL
  • NADH, 500μL

Colorimetric determination of aspartate transaminase activity.

  • Based on the quantification of oxaloacetate produced by AST
  • In this assay, oxaloacetate and NADH are converted to malate and NAD by the enzyme malate dehydrogenase
  • The decrease in NADH absorbance at 340nm is proportional to AST activity

Specifications

Calibration Range 2 to 100U/L
Content And Storage Store the kit at -20°C.
Description Aspartate Transaminase Assay Kit is a colorimetric determination of aspartate transaminase activity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 24mL assay buffer, 120μL Cofactors, 120μL enzyme mix, 500μL NADH
Quantity 1 Kit
Sample Type Biological Sample, Plasma, Serum
Sample Volume 200 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.