Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ATPase/GTPase Assay Kit

Catalog No. 89028342 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-342 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-342 Supplier Abnova™ Supplier No. KA1610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly sensitive method for determining ATPase/GTPase activities

  • Detection Method: Colorimetric

Functional Study

Specifications

Content And Storage Store the kit at 4°C.
Description ATPase/GTPase Assay Kit is a highly sensitive method for determining ATPase/GTPase activities.
Detection Method Colorimetric
Product Type Activity Detection Assay Kit
For Use With (Application) Functional Study
Kit Contents 50mL Reagent, 10mL assay buffer, 1mL standard 1mM Phosphate
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.