Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Autophagy/Cytotoxicity Dual Staining Kit

Catalog No. 89024589 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-589 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-589 Supplier Abnova™ Supplier No. KA1299
Add to Cart
Add to Cart

Fluorescence-labeled method for screening autophagy and cytotoxicity at the cellular level

Functional Study, Immunofluorescence

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Autophagy/Cytotoxicity Dual Staining Kit is a fluorescence-labeled method for screening autophagy and cytotoxicity at the cellular level.
Detection Method Fluorometric
For Use With (Application) Flow Cytometry, Functional Study, Immunofluorescence
Includes 100μL Monodansylcadaverine, 5 tablets buffer tablet, 250μL Propidium Iodide solution, 50μL Tamoxifen (100mM)
Quantity 1 Kit
Type Cytotoxicity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.