Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | beta2-Glycoprotein I Screen ELISA Kit is an indirect solid phase enzyme immunoassay for the semiquantitative screening of IgG, IgM and IgA class autoantibodies against b2-Glycoprotein I in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 1.5mL Controls with Anti-2 -Glycoprotein I Antibodies of IgG/IgM/IgA class in Serum/Buffer Matrix 4 vials, 20mL Sample Buffer 2 vials, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 15mL 96-well plate 1 plate wash solution 1 vial |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.