Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bilirubin Assay Kit

Catalog No. 89028346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-346 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-346 Supplier Abnova™ Supplier No. KA1614
Add to Cart
Add to Cart

Includes

  • Reagent A, 30mL
  • Reagent B, 10mL
  • Reagent C, 30mL
  • Saline, 50mL
  • Calibrator (equivalent to 5mg/dL bilirubin), 2mL

For quantitative colorimetric bilirubin determination at 530nm

Specifications

Content And Storage Store the kit at 4°C.
Description Bilirubin Assay Kit is a quantitative colorimetric bilirubin determination at 530 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 30mL Reagent A, 10mL reagent B, 30mL reagent C, 50mL Saline, 2mL calibrator (equivalent to 5mg/dL Bilirubin)
Quantity 1 Kit
Sample Volume 50 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.