Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Blue Cell Viability Assay Kit

Catalog No. 89028337 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-337 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-337 Supplier Abnova™ Supplier No. KA1605
Add to Cart
Add to Cart

Non-radioactive fluorescent assay for cell proliferation and cytotoxicity. Simply add a single reagent, the CellQuanti-Blue reagent, to the cell culture and measure the fluorescence intensity (excitation wavelength = 530 to 570nm, emission wavelength = 590 to 620nm) after an incubation step.

  • Reagent is compatible with all culture media and with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates
  • Applications include cell proliferation, cytotoxicity and apoptosis

Specifications

Content And Storage 4°C
Description Blue Cell Viability Assay Kit is a non-radioactive fluorescent assay for cell proliferation and cytotoxicity.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 50mL Reagent
Quantity 1 Kit
Type Cell Viability Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.