Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C Cell Division Assay Kit

Catalog No. 89024592 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-592 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-592 Supplier Abnova™ Supplier No. KA1302
Add to Cart
Add to Cart

Includes

  • CFSE Stock Solution 3 vials/100μL
  • Cell-Based Assay Buffer Tablet 4 tablets

Provides easy-to-use format for labeling and tracing cells through successive cell divisions which can be used to study the induction and inhibition of cell division in any in vitro model.

  • CFSE Cell Division Assay Kit is a fluorescence-based method for tracing cell divisions
  • The kit contains sufficient reagents for labeling and analyzing 100 cell samples by flow cytometry
  • CFSE can also be combined with any fluorochrome compatible with fluorescein for use in flow cytometry

Specifications

Content And Storage Store the kit at -20°C.
Description CFSE Cell Division Assay Kit is a fluorescence-based method for tracing cell divisions.
Detection Method Fluorometric
For Use With (Application) Flow Cytometry
Includes 100μL CFSE stock solution, 4 tablets buffer tablet
Quantity 1 Kit
Type Cell Division Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.