Learn More
Specifications
Specifications
| Calibration Range | 0.001 to 5μg/mL |
| Conjugate | HRP (Horseradish Peroxidase) |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Sample Type | Biological Fluids |
| Sample Volume | 50 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.