Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Calcium Assay Kit

Catalog No. 89028376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-376 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-376 Supplier Abnova™ Supplier No. KA1644
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A, 50mL
  • Reagent B, 50mL
  • Calcium standard: 20mg/dL Ca2+, 1mL

For quantitative colorimetric calcium determination at 612nm in saliva, serum, urine

Specifications

Calibration Range 0.08 to 20mg/dL
Content And Storage Store the kit at 4°C.
Description Calcium Assay Kit is a quantitative colorimetric calcium determination at 612 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent A, 50mL reagent B, 1mL calcium standard 20mg/dL Ca2
Quantity 1 Kit
Sample Type Saliva, Serum, Urine
Sample Volume 15 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.