Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Caspase-3 Assay Kit (Fluorometric)

Catalog No. 89101202 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-202 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-202 Supplier Abnova™ Supplier No. KA3786
Add to Cart
Add to Cart

Caspase-3 Assay Kit is a quantitative fluorimetric determination of caspase-3 at FL400/490 nm.

  • Safe, non-radioactive assay
  • Convenient and high-throughput
  • Homogeneous mix-incubate-measure type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Store at -20°C
  • For research use only

Specifications

Description Caspase-3 Assay Kit is a quantitative fluorimetric determination of caspase-3 at FL400/490 nm.
Quantity 1 Kit
Product Type Assay kit
Content And Storage Store the kit at -20°C
For Use With (Application) Functional Study
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.