Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Caspase-7 Immunoassay Kit

Catalog No. 89028026 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-026 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-026 Supplier Abnova™ Supplier No. KA0754

Caspase-7 Immunoassay Kit is used for specific, quantitative detection of caspase-7 activity.

  • Kit provides an effective immunosorbent enzyme assay for the specific, quantitative detection of caspase-7 activity in microtiter plates
  • Assay utilizes caspase-7 polyclonal antibody to capture activated caspase-7 from cell lysates
  • Simple, straightforward, and can be performed directly in microtiter plates
  • Free AFC can then be analyzed fluorometrically using a fluorescence plate reader
  • Kit has been configured for research use only, not to be used on humans
  • Store kit at -20°C (after thawing, store Cell Lysis Buffer, Coating Buffer, and Incubation Buffer at 4°C)
  • All reagents are stable for up to 6 months under proper storage conditions

Specifications

Description Caspase-7 Immunoassay Kit is used for specific, quantitative detection of caspase-7 activity.
Quantity 1 Kit
Content And Storage Store the kit at -20°C
For Use With (Application) Functional Study
Form Liquid (DMSO), Powder (DTT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.