Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Catalase Assay Kit

Catalog No. 89028358 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-358 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-358 Supplier Abnova™ Supplier No. KA1626
Add to Cart
Add to Cart

Includes

  • Assay buffer, 25mL
  • HRP enzyme, 120μL
  • Dye reagent, 120μL
  • H2O2 solution, 100μL (3% H2O2)
  • Positive control, 8μL catalase

For quantitative colorimetric/fluorometer catalase determination in cell culture, plasma, saliva, serum, urine

Specifications

Calibration Range 0.2 to 5U/L
Content And Storage Store the kit at -20°C.
Description Catalase Assay Kit is a quantitative colorimetric/fluoriemtric catalase determination.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 25mL assay buffer, 120μL HRP enzyme, 120μL dye reagent, 100μL 3% H2O2 solution, 8μL positive control Catalase
Quantity 1 Kit
Sample Type Cell Culture, Plasma, Saliva, Serum, Urine
Sample Volume 10 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.