Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL17 (Mouse) ELISA Kit

Catalog No. 89101098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-098 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-098 Supplier Abnova™ Supplier No. KA3686
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-mouse CCL17 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse CCL17 standard, 10ng x 2
  • Biotinylated anti-mouse CCL17 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Ccl17. Sandwich enzyme immunoassay.

Ccl17 (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 31.2 to 2000pg/mL
Content And Storage Store the kit at -20°C
Description Ccl17 (Mouse) is a sandwich enzyme immunoassay for the quantitative measurement of mouse Ccl17.
Conjugate Biotin
Cross Reactivity Mouse
Detection Method Colorimetric
Product Type Ccl17 (Mouse) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant mouse CCL17
Sufficient For 96 Assays
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.