Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL7 (Human) ELISA Kit

Catalog No. 89100788 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
15 to 800 pg/mL
15.6 to 1000 pg/mL
30 to 1600 pg/mL
Detection Limit:
1 pg/mL
10 pg/mL
30 pg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit
89-100-788 15 to 800 pg/mL 10 pg/mL
89-100-208 30 to 1600 pg/mL 30 pg/mL
89-101-008 15.6 to 1000 pg/mL 1 pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-788 Supplier Abnova™ Supplier No. KA3085
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-human MCP-3 antibody, 96 (8 x 12) wells
  • Standard (1080pg/mL), 0.5mL
  • Standard diluent buffer, 1.5mL
  • Wash buffer (30x), Dilution: 1:30, 20mL
  • Sample diluent buffer, 6mL
  • HRP conjugated anti-human MCP-3 antibody (RTU), 6mL
  • Stop solution, 6mL
  • TMB substrate A, 6mL
  • TMB substrate B, 6mL
  • Plate sealer, 2 slices
  • Hermetic bag, 1 bag

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15 to 800 pg/mL
Conjugate HRP
Cross Reactivity Human
Description CCL7 (Human) ELISA Kit is intended for the quantitative measurement of human CCL7.
Detection Limit 10 pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-human MCP-3 antibody (96 (8 x 12) wells), Standard (1080pg /mL): 0.5mL, Standard diluent buffer (1.5mL), Wash buffer (30x), Dilution: 1:30 (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human MCP-3 antibody (RTU): 6mL, Stop solution (6mL), TMB substrate A and B (6mL each), Plate sealer (2 slices), One hermetic bag
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type CCL7 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.