Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CD86 (Rat) ELISA Kit

Catalog No. 89101100 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-100 Supplier Abnova™ Supplier No. KA3688
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-rat CD86 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant rat CD86 standard, 10ng x 2
  • Biotinylated anti-rat CD86 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively rat Cd86. Sandwich enzyme immunoassay.

Cd86 (Rat) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 62.5 to 4000pg/mL
Content And Storage Store the kit at -20°C
Description Cd86 (Rat) is a sandwich enzyme immunoassay for the quantitative measurement of rat Cd86.
Conjugate Biotin
Cross Reactivity Rat
Detection Method Colorimetric
Product Type Cd86 (Rat) ELISA Kit
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and recombinant rat CD86
Sufficient For 96 Assays
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.