Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cell Cycle Phase Determination Kit

Catalog No. 89024591 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-591 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-591 Supplier Abnova™ Supplier No. KA1301
Add to Cart
Add to Cart

Includes

  • Propidium iodide reagent (powder), 1 vial/1mg
  • Cell-based assay buffer tablets, 2 tablets
  • RNase A solution, 1 vial/1mL
  • Cell cycle phase determination fixative, 1 vial/100mL

Used for determining cell cycle phase and flow cytometry

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Cell Cycle Phase Determination Kit is a fluorescence-labeled method for determining cell cycle phase.
Detection Method Fluorometric
For Use With (Application) Flow Cytometry
Includes 1mg Propidium Iodide reagent powder, 2 tablets buffer tablets, 1mL RNase A solution, 100mL cell cycle phase determination fixative
Quantity 1 Kit
Type Cell Cycle Phase Determination Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.