Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cellular UV DNA-Damage Detection Kit

Catalog No. 89101136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-136 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-136 Supplier Abnova™ Supplier No. KA0293
Add to Cart
Add to Cart

A non-isotopic immunoassay used for the semi-quantitative measurement of CPDs (cyclobutane pyrimidine dimers) in genomic DNA formed by UV-irradiation to cells.

Applications include:
  • Detection and semi-quantification of cyclobutane pyrimidine dimers in cells.
  • Monitoring the effects of UV on formation of cyclobutane pyrimidine dimers in cells.
  • Monitoring the effects of UV protection reagent on formation of cyclobutane pyrimidine dimers in cells.
  • Study on the repair mechanisms of UV-induced DNA damage by the nucleotide excision repair in cells
Principle of the Assay. The Abnova Cellular UV DNA-Damage Detection Kit is based on the formation of CPDs on the genomic DNA after UV-irradiation in the cells that are cultured in microtiter plates. After formation of CPDs on DNA, CPDs in the cell is detected by anti-CPDs antibody. During UV-irradiation is carried out on wells of the microtiter plate, CPDs will be formed on the DNA of living cells. To enable antibody binding to the CPDs on the genomic DNA, cells must be fixed, permeabilized and the DNA denatured. Detector anti-CPDs antibody is pipetted into the wells and allowed to incubate for one hour, during which time it binds to any CPDs. Unbound antibody is washed away and horseradish peroxidase-conjugated secondary antibody is added, which binds to the detector antibody. The horseradish peroxidase catalyzes the conversion of the chromogenic substrate tetra-methylbenzidine (TMB) from a colorless solution to a blue solution (or yellow after the addition of stopping reagent). The color is quantified by spectrophotometry and reflects the relative amount of CPDs in the cells.

For research use only; not for use in diagnostic or therapeutic procedures.

Specifications

Content And Storage Store the kit at 4°C.
Description Cellular UV DNA-Damage Detection Kit is a non-isotopic immunoassay used for the semi-quantitative measurement of CPDs (cyclobutane pyrimidine dimers) in genomic DNA formed by UV-irradiation to cells.
For Use With (Application) Functional Study
Includes 200 assays
Quantity 1 Kit
Sample Type Genomic DNA
Sample Volume 1-4 x 10^4 cells/well
Type Detection Assay Kit
Regulatory Status RUO
Species Human, Bovine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.