Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Store the kit at 4°C. |
| Description | CETP Activity Assay Kit is used to determining of the presence of CETP. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 1mL Donor molecule, 1mL acceptor molecule, 5mL CETP assay buffer 10X, 30μL positive control (Rabbit Serum) |
| Quantity | 1 Kit |
| Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
| Type | Activity Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?