Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Chloride Assay Kit

Catalog No. 89028377 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-377 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-377 Supplier Abnova™ Supplier No. KA1645
Add to Cart
Add to Cart

Includes

  • Reagent, 50mL
  • Chloride standard, (35mg/dL Cl-), 1mL

For quantitative colorimetric chloride determination at 610nm in plasma, saliva, serum, sweat, urine

Specifications

Calibration Range 0.2 to 10mm
Content And Storage Store the kit at 4°C.
Description Chloride Assay Kit is a quantitative colorimetric chloride determination at 610 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent, 1mL Chloride standard (35mg/dL Cl-)
Quantity 1 Kit
Sample Type Plasma, Saliva, Serum, Sweat, Urine
Sample Volume 10 μL
Type Cellular Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.