Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cholesterol Assay Kit

Catalog No. 89024593 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-593 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-593 Supplier Abnova™ Supplier No. KA1303
Add to Cart
Add to Cart

Provides a simple fluorometric method for the sensitive quantitation of cholesterol in serum and plasma. The cholesterol assay is based on an enzyme-coupled reaction that detects both free cholesterol and cholesteryl esters.

Specifications

Content And Storage Store the kit at -20°C.
Description Cholesterol Assay Kit is a fluorescence-labeled method for quantification of cholesterol.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 3mL Cholesterol assay buffer 10X, 100μL Cholesterol assay standard, 2 vials Cholesterol assay detector, 1 vial Cholesterol assay Horseradish Peroxidase, 1 vial Cholesterol assay Oxidase, 1 vial Cholesterol assay Esterase, 500μL
Quantity 1 Kit
Type Cellular Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.