Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cholesterol Cell-Based Detection Assay Kit

Catalog No. 89024594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-594 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-594 Supplier Abnova™ Supplier No. KA1304
Add to Cart
Add to Cart

Includes

  • Cell-based assay fixative, 2 vials
  • Cholesterol detection wash buffer, 1 vial
  • Cholesterol detection filipin III, 1 vial
  • Cholesterol detection assay buffer, 1 vial
  • Cell-based assay U18666A, 1 vial

Used for histochemical identification of unesterified cholesterol in vivo and in-vitro, immunofluorescence and functional study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Cholesterol Cell-Based Detection Assay Kit is a fluorescence-labeled method for histochemical identification of unesterified cholesterol in vivo and in vitro.
Detection Method Fluorometric
For Use With (Application) Functional Study, Immunofluorescence
Includes 2 vials fixative, 1 vial Cholesterol detection wash buffer, 1 vial Cholesterol detection Filipin III, 1 vial Cholesterol detection assay buffer, 1 vial U18666A
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.