Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Choline Assay Kit

Catalog No. 89028394 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-394 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-394 Supplier Abnova™ Supplier No. KA1662
Add to Cart
Add to Cart

For quantitative colorimetric/fluorometric choline determination in cell culture, milk, plasma, saliva, serum, tissue, urine

Specifications

Calibration Range 1 to 100μM colorimetric assay, 0.2 to 10μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Choline Assay Kit is a quantitative colorimetric/fluorometric choline determination.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 10mL assay buffer, enzyme mix (Dried) 1 vial, 120μL dye reagent, 400μL standard 2mM Choline
Quantity 1 Kit
Sample Type Cell Culture, Milk, Plasma, Saliva, Serum, Tissue, Urine
Sample Volume 20 μL
Type Cellular Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.