Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Clu (Mouse), Biotin, ELISA Kit

Catalog No. 89101034 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-034 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-034 Supplier Abnova™ Supplier No. KA3355
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti- mouse Clusterin antibody 96 (8x12) wells
  • Lyophilized recombinant mouse Clusterin standard 20ng x 2
  • Biotinylated anti- mouse Clusterin antibody(dilution 1:100) 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100) 130μL
  • Sample diluent buffer 30mL
  • antibody diluent buffer 12mL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl5

Specifications

Calibration Range 312 to 20000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Clu (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Clu.
Detection Limit 10 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernate, plasma (EDTA, Heparin) and serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type CLU (Mouse) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.