Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Cobalt reagent, 1mLl
- Cobalt chloride standard (1μmol), lyophilized, 1 vial
Specifications
Specifications
| Calibration Range | 3 to 50nmol |
| Content And Storage | Store the kit at room temperature. |
| Description | Cobalt Assay Kit is used for quantitating Co+2 in a variety of samples. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 1mL Cobalt reagent, 1 vial Cobalt Chloride standard (1μM) (lyophilized) |
| Quantity | 1 Kit |
| Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
| Type | Cellular Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?