Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ COL18A1 (Human) ELISA Kit

Catalog No. 89028444 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Human Milk, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine
Cell Lysates, Plasma, Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-444 CCS, Plasma, Serum, Urine
89-100-478 Cell Lysates, Plasma, Serum
89-101-057 CCS, Human Milk, Plasma (EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-028-444 Supplier Abnova™ Supplier No. KA1708

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human COL18A1

Specifications

Conjugate Biotin
Cross Reactivity Human
Description COL18A1 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human COL18A1.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB One-Step Substrate reagent, 8mL stop solut
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store the kit at 4°C. For best results, store the components as described in the protocol.
For Use With (Application) Quantification
Product Type COL18A1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.