Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ COMP (Human) ELISA Kit

Catalog No. 89100009 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
156 to 10000pg/mL
4 to 128ng/mL
Detection Limit:
0.4ng/mL
10pg/mL
Conjugate:
Biotin
HRP
Content And Storage:
Store the kit at -20°C
Store the kit at 4°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit Conjugate Content And Storage
89-100-009 4 to 128ng/mL 0.4ng/mL HRP Store the kit at 4°C
89-101-021 156 to 10000pg/mL 10pg/mL Biotin Store the kit at -20°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-009 Supplier Abnova™ Supplier No. KA0021
Add to Cart
Add to Cart

Includes

  • Antibody Coated Microtiter Strips, 96 wells
  • Biotin Labelled Antibody, 13mL
  • Streptavidin-HRP Conjugate, 13mL
  • Master Standard, 2 vials
  • Quality Control HIGH, 2 vials
  • Quality Control LOW, 2 vials
  • Dilution Buffer, 2 x 13mL
  • Wash Solution Conc. (10x), 100mL
  • Substrate Solution, 13mL
  • Stop Solution, 13mL

  • The kit measures COMP in serum and plasma (EDTA, citrate, heparin)
  • Quality Controls are human serum based
  • Standard is recombinant protein based
  • Components of the kit are provided ready to use, concentrated or lyophilized

Specifications

Calibration Range 4 to 128ng/mL
Conjugate HRP
Cross Reactivity Human
Description COMP (Human) ELISA Kit is a double monoclonal sandwich enzyme immunoassay for the quantitative measurement of human COMP in serum or plasma.
Detection Limit 0.4ng/mL
Detection Method Colorimetric
Includes Antibody Coated Microtiter Strips (96 wells), Biotin Labelled Antibody (13mL), Streptavidin-HRP Conjugate (13mL), Master Standard (2 vials), Quality Control HIGH (2 vials), Quality Control LOW (2 vials), Dilution Buffer (2 x 13mL), Wash Solution Conc. 10x (100mL), Substrate Solution (13mL), Stop Solution (13mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity 0.4 ng/mL
Total Assay Time <3.5 hr.
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type COMP (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.