Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Copper Assay Kit

Catalog No. 89028347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-347 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-347 Supplier Abnova™ Supplier No. KA1615
Add to Cart
Add to Cart

Includes

  • Reagent A, 10mL
  • Reagent B, 1.5mL
  • Reagent C, 40mL
  • Copper standard: 1.5mg/dL Cu2+, 1mL

Designed to measure copper with no or minimal sample treatment. The intensity of the color, measured at 359nm, is directly proportional to copper concentration in the sample.

  • Uses a chromogen that forms a colored complex specifically with copper ions
  • The optimized formulation substantially reduces interference by substances in the raw samples

Specifications

Calibration Range 1 to 47μM
Content And Storage Store the kit at 4°C.
Description Copper Assay Kit is a quantitative colorimetric copper determination at 359 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL Reagent A, 1.5mL reagent B, 40mL reagent C, 1mL Copper standard 1.5mg/dL Cu2+
Quantity 1 Kit
Sample Type Biological Sample, Beverage Sample, Environmental Sample, Food Sample
Sample Volume 100 μL
Type Cellular Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.