Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol ELISA Kit

Catalog No. 89028717 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Dried Fecal Extract, Plasma, Saliva, Serum, Tissue Culture Medium Sample, Urine
Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-717 Urine
89-100-919 Dried Fecal Extract, Plasma, Saliva, Serum, Tissue Culture Medium Sample, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-717 Supplier Abnova™ Supplier No. KA1901
Add to Cart
Add to Cart

For the quantitative measurement of free Cortisol in Urine

Specifications

Conjugate HRP
Description Cortisol ELISA Kit is an enzyme immunoassay for the quantitative measurement of free cortisol in urine.
Detection Limit 2ng/mL
Detection Method Colorimetric
Includes 1mL Standard A, 1mL standard B, 1mL standard C, 1mL standard D, 1mL standard E, 1mL Control 1, 1mL Control 2, 100mL INC Buffer, 1mL Conjugated-Conc, 96-well microplate, 15mL Substrate, 15mL stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Urine
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.