Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Creatine Assay Kit

Catalog No. 89028398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-398 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-398 Supplier Abnova™ Supplier No. KA1666
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay buffer, 20mL
  • Enzyme A, 120μL
  • Enzyme B, 220μL
  • Standard: 20mM creatine, 400μL
  • Dye reagent, 220μL

Colorimetric/fluorometer determination of creatine in plasma, saliva, serum, urine

Specifications

Calibration Range 4 to 1000μM colorimetric assay, 0.5 to 50μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Creatine Assay Kit is a quantitative colorimetric/fluorimetric determination of creatine.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 20mL assay buffer, 120μL enzyme A, 220μL enzyme B, 400μL standard 20mM Creatine, 220μL dye reagent
Quantity 1 Kit
Sample Type Plasma, Saliva, Serum, Urine
Sample Volume 10 μL
Type Cellular Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.