Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CST3 (Human) ELISA Kit

Catalog No. 89100379 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-379 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-100-008 CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
89-100-921 Plasma (EDTA, Heparin), Serum, Tissue Culture Medium, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-379 Supplier Abnova™ Supplier No. KA2107
Add to Cart
Add to Cart

Includes

  • Lyophilized recombinant human Cystatin C standard: 20ng/tube x 2
  • One 96-well plate precoated with anti- human Cystatin C antibody
  • Sample diluent buffer: 30mL
  • Biotinylated anti-human Cystatin C antibody: 130μL, dilution 1:100
  • Antibody diluent buffer: 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC): 130μL, dilution 1:100
  • ABC diluent buffer: 12mL
  • TMB color developing agent: 10mL
  • TMB stop solution: 10mL

CST3 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 0.312 to 20ng/mL
Conjugate Biotin
Cross Reactivity Human
Description CST3 (Human ) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CST3.
Detection Limit <10 pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Lyophilized recombinant Human CST3 standard (10ng x 2 vials), Sample diluent buffer (30mL), Biotinylated anti-Human CST3 antibody (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase complex (ABC): 130μL, ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type CST3 (Human) ELISA Kit
Specificity No detectable cross-reactivity with any other cytokine
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.