Learn More
Description
Includes
- BLC Microplate (Item A): coated with anti-human BLC, 96-wells (12 strips x 8 wells)
- Wash Buffer Concentrate (20x) (Item B): 20x concentrated solution, 25mL
- Standard recombinant human BLC (Item C), 2 vials
- Assay Diluent A (Item D): 0.09% sodium azide as preservative, For Standard/Sample (serum/plasma) diluent, 30mL
- Assay Diluent B (Item E): 5x concentrated buffer, For Standard/Sample (cell culture medium/urine) diluent, 15mL
- Detection Antibody BLC (Item F): Biotinylated anti-human BLC (each vial is enough to assay half microplate), 2 vials
- HRP-Streptavidin Concentrate (Item G): 200x concentrated, HRP-conjugated Streptavidin, 200μL
- TMB One-Step Substrate Reagent (Item H): 3, 3', 5, 5'-tetramethylbenzidine (TMB) in buffered solution, 12mL
- Stop Solution (Item I): 0.2M sulfuric acid, 8mL
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Description | CXCL13 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CXCL13. |
| Detection Limit | 1.5pg/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB One-Step Substrate reagent, 8mL stop solut |
| Quantity | 96 Assays |
| Sample Type | Cell Culture Supernatant, Plasma, Serum, Urine |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.