Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNA Assay Kit

Catalog No. 89028371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-371 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-371 Supplier Abnova™ Supplier No. KA1639
Add to Cart
Add to Cart

Fluorometer quantitation of nanogram DNA

Specifications

Calibration Range 2 to 40ng
Content And Storage Store DNA Standard at -20°C. Store Reagent at 4°C.
Description DNA Assay Kit is a fluorimetric quantitation of nanogram DNA.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 50mL Reagent, 1mL 10μg/mL standard, Calf Thymus DNA
Quantity 1 Kit
Type DNA Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.