Learn More
Abnova™ DNA Methyltransferase 1 Activity/Inhibitor Screening Assay Core Kit
Catalog No. 89028410 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-028-410 1 Kit
1 options
Catalog No. 89-028-410 Supplier Abnova™ Supplier No. KA1604
Add to Cart
Add to Cart
Used for screening DNMT 1 inhibitors
Specifications
Content And Storage Store MO3,and MO6, at -20°C away from light. Store MO1, MO2, MO5, MO7, and 8-Well Substrate-Coated Strips at 4°C. Store MO8 at room temperature.
Description DNA Methyltransferase 1 Activity/Inhibitor Screening Assay Core Kit is used for screening Dnmt 1 inhibitors.
For Use With (Application) Functional Study
Includes 11mL MO1 (wash buffer 10X), 2mL MO2 (DNMT assay buffer), 35μL MO3 (Adomet, 8mM), 5μL MO5 (Capture antibody), 10μL MO6 (detection antibody), 6mL MO7 (developing solution), 3mL MO8 (stop solution), 6
Quantity 1 Kit
Type Activity/inhibition Assay Kit