Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNA Methyltransferase 3B Activity/Inhibitor Screening Assay Core Kit

Catalog No. 89028241 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-241 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-241 Supplier Abnova™ Supplier No. KA1547
Add to Cart
Add to Cart

Used for screening DNA methyltransferase 3B inhibitors

Specifications

Content And Storage Store M13, M14, and M16 at -20°C. Store MT1, MT2, MT5, MT7 and 8-well substrate-coated strips at 4°C away from light. Store MT8 at room temperature.
Description DNA Methyltransferase 3B Activity/Inhibitor Screening Assay Core Kit is use for screening DNA methyltransferase 3B inhibitors.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 11mL MT1 (wash buffer 10X), 2mL MT2 (DNMT assay buffer), 35μL MT3 (adomet, 8mM), 25μL MT4 (active DNMT3B, 2 KU/mL), 5μL MT5 (Capture antibody), 10μL MT6 (detection antibody, 6mL MT7 (
Quantity 1 Kit
Type Activity/inhibition Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.