Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNMT Activity/Inhibition Assay Ultra Kit (Fluorometric)

Catalog No. 89028211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-211 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-211 Supplier Abnova™ Supplier No. KA1516
Add to Cart
Add to Cart

For screening DNMT inhibitors

  • Sample Type: Nuclear Extract, Purified Enzyme
  • Sample Volume: 1-5μL (5 to 10μg nuclear extract or 10 to 100ng purified enzyme included)

Functional Study

Specifications

Calibration Range 0.2 to 10μg nuclear extract, 0.2 to 200ng purified Enzyme
Content And Storage Store MK3, MK4, MK6, MK7 and MK8 at -20°C away from light. Store MK1, MK5, MK9 and 8-Well Assay Strips at 4°C away from light. Store all other components at room temperature away from light.
Description DNMT Activity/Inhibition Assay Ultra Kit (Fluorometric) is use for screening DNMT inhibitors.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 14mL MK1 (wash buffer 10X), 4mL MK2 (DNMT assay buffer), 60μL MK3 (Adomet, 50X), 6μL MK4 (DNMT enzyme control, 50μg/mL), 5μL MK5 (Capture antibody, 1000μg/mL), 6
Quantity 1 Kit
Sample Type Nuclear Extract, Purified Enzyme
Sample Volume 1 to 5 μL
Type Activity/inhibition Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.